Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGGCCCTTACCTGTAGGGGGCGC[C/T]TGGGAGCGGGGCCTTGGCAGTTCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604528 MIM: 604037 | ||||||||||||||||||||
Literature Links: |
KCNH4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCNH4 - potassium voltage-gated channel subfamily H member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012285.2 | 2904 | Silent Mutation | CAA,CAG | Q,Q 857 | NP_036417.1 | |
XM_017024400.1 | 2904 | Silent Mutation | CAA,CAG | Q,Q 857 | XP_016879889.1 | |
XM_017024401.1 | 2904 | Silent Mutation | CAA,CAG | Q,Q 808 | XP_016879890.1 | |
XM_017024402.1 | 2904 | Silent Mutation | CAA,CAG | Q,Q 797 | XP_016879891.1 | |
XM_017024403.1 | 2904 | Intron | XP_016879892.1 |
RAB5C - RAB5C, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |