Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGCGATGCGGTTGAATGCTTTCC[C/T]ACATTCATTACATTCATAGGGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF624 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF624 - zinc finger protein 624 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020787.3 | 1608 | Missense Mutation | AGG,GGG | R,G 506 | NP_065838.2 | |
XM_006721562.3 | 1608 | Missense Mutation | AGG,GGG | R,G 506 | XP_006721625.1 | |
XM_011523970.2 | 1608 | Missense Mutation | AGG,GGG | R,G 504 | XP_011522272.1 |