Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGTCTGGTTAGGGAGAGCTCCGG[A/G]GGAGAGGTCCGAAAGCAGCAGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 155120 MIM: 611661 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAM11 - ADAM metallopeptidase domain 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318933.1 | 116 | UTR 5 | NP_001305862.1 | |||
NM_002390.5 | 116 | Silent Mutation | GGA,GGG | G,G 59 | NP_002381.2 | |
XM_005257373.3 | 116 | Silent Mutation | GGA,GGG | G,G 59 | XP_005257430.1 | |
XM_017024647.1 | 116 | UTR 5 | XP_016880136.1 |
DBF4B - DBF4 zinc finger B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |