Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACTCAGAGCACGCCCCTGTCTGT[C/T]GGGTAGGTGGGTTACTTGCTCTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAHCC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BAHCC1 - BAH domain and coiled-coil containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291324.1 | Intron | NP_001278253.1 | ||||
XM_011525063.2 | Intron | XP_011523365.1 | ||||
XM_017024898.1 | Intron | XP_016880387.1 | ||||
XM_017024899.1 | Intron | XP_016880388.1 |
MIR4740 - microRNA 4740 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |