Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTAGTATAAAGGTTTTCTAAAA[C/G]GATTGATTCTCATTCTGAGGCAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605468 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC42EP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDC42EP4 - CDC42 effector protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012121.4 | 2423 | UTR 3 | NP_036253.2 | |||
XM_005257182.2 | 2423 | UTR 3 | XP_005257239.1 |