Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGGGTAAGAATTGGAAGAATTGT[A/G]TAGGAGGCAGTAGTCAAAAAGTAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610304 MIM: 609178 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DERL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DERL2 - derlin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIS12 - MIS12, kinetochore complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258217.1 | 243 | UTR 5 | NP_001245146.1 | |||
NM_001258218.1 | 243 | Intron | NP_001245147.1 | |||
NM_001258219.1 | 243 | UTR 5 | NP_001245148.1 | |||
NM_001258220.1 | 243 | Intron | NP_001245149.1 | |||
NM_024039.2 | 243 | UTR 5 | NP_076944.1 | |||
XM_005256797.2 | 243 | UTR 5 | XP_005256854.1 | |||
XM_017025033.1 | 243 | UTR 5 | XP_016880522.1 | |||
XM_017025034.1 | 243 | UTR 5 | XP_016880523.1 | |||
XM_017025035.1 | 243 | UTR 5 | XP_016880524.1 |