Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTGACATCATAGACCGAGAAGC[A/C]GATGGAAGTGACAGTTTGGAGGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605785 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLEKHH3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLEKHH3 - pleckstrin homology, MyTH4 and FERM domain containing H3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TUBG2 - tubulin gamma 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320509.1 | 629 | Silent Mutation | GCA,GCC | A,A 126 | NP_001307438.1 | |
NM_016437.2 | 629 | Silent Mutation | GCA,GCC | A,A 126 | NP_057521.1 | |
XM_011524614.2 | 629 | UTR 5 | XP_011522916.1 | |||
XM_017024453.1 | 629 | UTR 5 | XP_016879942.1 | |||
XM_017024454.1 | 629 | UTR 5 | XP_016879943.1 | |||
XM_017024455.1 | 629 | UTR 5 | XP_016879944.1 | |||
XM_017024456.1 | 629 | UTR 5 | XP_016879945.1 | |||
XM_017024457.1 | 629 | UTR 5 | XP_016879946.1 |