Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGCATCACCAGCCGGTCATCCAC[A/G]AAGTAGAAGCTGTCAGACTGGGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600838 MIM: 610271 MIM: 604011 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOXN1 - forkhead box N1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGS - phosphatidylinositol glycan anchor biosynthesis class S | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UNC119 - unc-119 lipid binding chaperone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005148.3 | 1001 | Silent Mutation | TTC,TTT | F,F 221 | NP_005139.1 | |
NM_054035.2 | 1001 | UTR 3 | NP_473376.1 | |||
XM_011525459.2 | 1001 | Intron | XP_011523761.1 | |||
XM_017025289.1 | 1001 | Silent Mutation | TTC,TTT | F,F 126 | XP_016880778.1 |