Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCTGGCAGAACTCTGCGTGAGA[A/C]GAGGACAGCAAAAGCCCAGCCCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138254 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIN2C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRIN2C - glutamate ionotropic receptor NMDA type subunit 2C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000835.4 | 5350 | UTR 3 | NP_000826.2 | |||
NM_001278553.1 | 5350 | Intron | NP_001265482.1 | |||
XM_006721845.3 | 5350 | UTR 3 | XP_006721908.1 | |||
XM_006721846.3 | 5350 | UTR 3 | XP_006721909.1 | |||
XM_011524686.2 | 5350 | UTR 3 | XP_011522988.1 | |||
XM_011524687.2 | 5350 | UTR 3 | XP_011522989.1 | |||
XM_011524688.2 | 5350 | UTR 3 | XP_011522990.1 | |||
XM_011524689.2 | 5350 | UTR 3 | XP_011522991.1 | |||
XM_011524692.2 | 5350 | Intron | XP_011522994.1 | |||
XM_017024544.1 | 5350 | UTR 3 | XP_016880033.1 |
TMEM104 - transmembrane protein 104 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |