Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTTAACGATACTCCCATTTAAGA[C/T]TGCCTGTTTGGTTACTGTTTCTACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603111 | ||||||||||||||||||||
Literature Links: |
KRT222 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRT222 - keratin 222 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152349.2 | 672 | Missense Mutation | ATC,GTC | I,V 203 | NP_689562.1 |
SMARCE1 - SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily e, member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |