Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGTAGGTGCGGAACCACATGGCA[C/G]TGCCCGAGTTGGCCTGGTACTTGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605752 MIM: 607828 | ||||||||||||||||||||
Literature Links: |
HID1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HID1 - HID1 domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030630.2 | 2402 | Missense Mutation | ACT,AGT | T,S 753 | NP_085133.1 | |
XM_005257226.1 | 2402 | Missense Mutation | ACT,AGT | T,S 752 | XP_005257283.1 |
OTOP3 - otopetrin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |