Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGTAGCAGGAGAGCTCGGTACCC[C/G]CTGTAGCAGCCATCAACTGAATCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616334 | ||||||||||||||||||||
Literature Links: |
TMEM100 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM100 - transmembrane protein 100 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099640.1 | 2306 | Missense Mutation | GCG,GGG | A,G 47 | NP_001093110.1 | |
NM_018286.2 | 2306 | Missense Mutation | GCG,GGG | A,G 47 | NP_060756.2 | |
XM_017024814.1 | 2306 | Missense Mutation | GCG,GGG | A,G 47 | XP_016880303.1 | |
XM_017024815.1 | 2306 | Missense Mutation | GCG,GGG | A,G 47 | XP_016880304.1 | |
XM_017024816.1 | 2306 | Missense Mutation | GCG,GGG | A,G 47 | XP_016880305.1 |