Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGAGCAGCACGGTGGGAAAGCAGG[C/T]GTTTTCTTTTGGCCTACTTTGGTGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601574 | ||||||||||||||||||||
Literature Links: |
HEATR9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HEATR9 - HEAT repeat containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321395.1 | 1691 | Silent Mutation | ACA,ACG | T,T 493 | NP_001308324.1 | |
NM_152781.3 | 1691 | Silent Mutation | ACA,ACG | T,T 533 | NP_689994.2 | |
XM_005278233.2 | 1691 | Silent Mutation | ACA,ACG | T,T 499 | XP_005278290.1 | |
XM_005278235.1 | 1691 | Silent Mutation | ACA,ACG | T,T 459 | XP_005278292.1 | |
XM_006721802.3 | 1691 | Silent Mutation | ACA,ACG | T,T 351 | XP_006721865.1 | |
XM_011524586.2 | 1691 | Silent Mutation | ACA,ACG | T,T 499 | XP_011522888.1 | |
XM_017024424.1 | 1691 | Intron | XP_016879913.1 |
TAF15 - TATA-box binding protein associated factor 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |