Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGCCCTGAATGTGGTGTTCCTG[G/T]GTGGCGCCTTCATCTGCAGCATGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605752 MIM: 607827 MIM: 607828 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HID1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HID1 - HID1 domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OTOP2 - otopetrin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OTOP3 - otopetrin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272005.1 | 370 | Missense Mutation | GGT,TGT | G,C 84 | NP_001258934.1 | |
NM_178233.2 | 370 | Missense Mutation | GGT,TGT | G,C 102 | NP_839947.1 | |
XM_011524744.2 | 370 | Missense Mutation | GGT,TGT | G,C 73 | XP_011523046.1 |