Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTCCACAAACCCTCCGTGCACT[C/T]AGCCGAAAAGAACTGTGCCTATTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610730 MIM: 600940 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCT6B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCT6B - chaperonin containing TCP1 subunit 6B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LIG3 - DNA ligase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002311.4 | 177 | Silent Mutation | CTC,CTT | L,L 16 | NP_002302.2 | |
NM_013975.3 | 177 | Silent Mutation | CTC,CTT | L,L 16 | NP_039269.2 | |
XM_005257970.3 | 177 | Silent Mutation | CTC,CTT | L,L 25 | XP_005258027.1 | |
XM_006721896.3 | 177 | Silent Mutation | CTC,CTT | L,L 25 | XP_006721959.1 | |
XM_017024624.1 | 177 | Silent Mutation | CTC,CTT | L,L 16 | XP_016880113.1 |