Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCATCGAAAAAGGGAAACACCC[A/G]AAAGTTACAAAACAGTGGACAGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615514 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK12 - cyclin dependent kinase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015083.2 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | NP_055898.1 | |
NM_016507.3 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | NP_057591.2 | |
XM_005257456.3 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_005257513.1 | |
XM_005257458.4 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_005257515.1 | |
XM_011524892.2 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_011523194.1 | |
XM_011524893.2 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_011523195.1 | |
XM_011524894.2 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_011523196.1 | |
XM_011524895.2 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_011523197.1 | |
XM_011524896.2 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_011523198.1 | |
XM_011524897.2 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_011523199.1 | |
XM_011524898.2 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_011523200.1 | |
XM_011524899.2 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_011523201.1 | |
XM_011524900.2 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_011523202.1 | |
XM_011524901.2 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_011523203.1 | |
XM_011524902.2 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_011523204.1 | |
XM_011524903.2 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_011523205.1 | |
XM_011524905.2 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_011523207.1 | |
XM_011524906.2 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_011523208.1 | |
XM_011524907.2 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_011523209.1 | |
XM_017024744.1 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_016880233.1 | |
XM_017024745.1 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_016880234.1 | |
XM_017024746.1 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_016880235.1 | |
XM_017024747.1 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_016880236.1 | |
XM_017024748.1 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_016880237.1 | |
XM_017024749.1 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_016880238.1 | |
XM_017024750.1 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_016880239.1 | |
XM_017024751.1 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_016880240.1 | |
XM_017024752.1 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_016880241.1 | |
XM_017024753.1 | 1183 | Missense Mutation | AAA,GAA | K,E 208 | XP_016880242.1 |