Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGGGCCGGTCACTACCTTTTCTG[C/T]CTGACAAGTGTACATAAAACAATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600365 MIM: 607251 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABR - active BCR-related | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001092.4 | 3066 | UTR 3 | NP_001083.2 | |||
NM_001159746.2 | 3066 | UTR 3 | NP_001153218.1 | |||
NM_001256847.2 | 3066 | UTR 3 | NP_001243776.1 | |||
NM_001282149.1 | 3066 | UTR 3 | NP_001269078.1 | |||
NM_001322840.1 | 3066 | UTR 3 | NP_001309769.1 | |||
NM_001322841.1 | 3066 | UTR 3 | NP_001309770.1 | |||
NM_001322842.1 | 3066 | Intron | NP_001309771.1 | |||
NM_021962.4 | 3066 | UTR 3 | NP_068781.2 | |||
XM_011523810.2 | 3066 | UTR 3 | XP_011522112.1 | |||
XM_011523811.2 | 3066 | Intron | XP_011522113.2 | |||
XM_011523812.2 | 3066 | UTR 3 | XP_011522114.1 | |||
XM_011523813.2 | 3066 | UTR 3 | XP_011522115.1 | |||
XM_011523814.2 | 3066 | UTR 3 | XP_011522116.1 | |||
XM_011523815.2 | 3066 | UTR 3 | XP_011522117.1 | |||
XM_017024539.1 | 3066 | UTR 3 | XP_016880028.1 | |||
XM_017024540.1 | 3066 | UTR 3 | XP_016880029.1 | |||
XM_017024541.1 | 3066 | Intron | XP_016880030.1 | |||
XM_017024542.1 | 3066 | UTR 3 | XP_016880031.1 | |||
XM_017024543.1 | 3066 | UTR 3 | XP_016880032.1 |
TIMM22 - translocase of inner mitochondrial membrane 22 homolog (yeast) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |