Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGACTGGGCTGGGAGATAGAAATG[A/G]GGAAAGAGAGTTTGGTTACTGGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603330 MIM: 194524 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAH9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAH9 - dynein axonemal heavy chain 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF18 - zinc finger protein 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303281.1 | 2278 | UTR 3 | NP_001290210.1 | |||
NM_001303282.1 | 2278 | UTR 3 | NP_001290211.1 | |||
NM_144680.3 | 2278 | UTR 3 | NP_653281.2 | |||
XM_011524002.2 | 2278 | UTR 3 | XP_011522304.1 | |||
XM_017025006.1 | 2278 | UTR 3 | XP_016880495.1 | |||
XM_017025007.1 | 2278 | UTR 3 | XP_016880496.1 | |||
XM_017025008.1 | 2278 | UTR 3 | XP_016880497.1 | |||
XM_017025009.1 | 2278 | UTR 3 | XP_016880498.1 | |||
XM_017025010.1 | 2278 | UTR 3 | XP_016880499.1 | |||
XM_017025011.1 | 2278 | UTR 3 | XP_016880500.1 |