Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCTGCCCCTGAGAGCCGTCAAC[A/C]TCAACCTCCGCGCAGGGCCCTCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608921 | ||||||||||||||||||||
Literature Links: |
FAM64A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM64A - family with sequence similarity 64 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195228.1 | 400 | Intron | NP_001182157.1 | |||
NM_019013.2 | 400 | Intron | NP_061886.2 | |||
XM_011523944.2 | 400 | Missense Mutation | ATC,CTC | I,L 61 | XP_011522246.1 | |
XM_017024778.1 | 400 | Intron | XP_016880267.1 |
PITPNM3 - PITPNM family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |