Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGGAACCGGGTGCTCACCTCGTG[A/G]TTGGCGTGGATCTGGCGGAGGTACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611301 MIM: 607643 | ||||||||||||||||||||
Literature Links: |
FAAP100 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAAP100 - Fanconi anemia core complex associated protein 100 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025161.5 | 2581 | Silent Mutation | AAC,AAT | N,N 836 | NP_079437.5 | |
XM_006722111.2 | 2581 | Silent Mutation | AAC,AAT | N,N 698 | XP_006722174.1 | |
XM_006722112.3 | 2581 | Silent Mutation | AAC,AAT | N,N 685 | XP_006722175.1 | |
XM_017025162.1 | 2581 | Silent Mutation | AAC,AAT | N,N 685 | XP_016880651.1 | |
XM_017025163.1 | 2581 | Silent Mutation | AAC,AAT | N,N 685 | XP_016880652.1 |
FSCN2 - fascin actin-bundling protein 2, retinal | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |