Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCAGGTTCGGAGATGATCGAGAC[A/G]CAGGAGGACATCTACGTGGGCTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601964 MIM: 604497 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJC7 - DnaJ heat shock protein family (Hsp40) member C7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NKIRAS2 - NFKB inhibitor interacting Ras like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001349.2 | 179 | Silent Mutation | ACA,ACG | T,T 38 | NP_001001349.1 | |
NM_001144927.1 | 179 | Silent Mutation | ACA,ACG | T,T 38 | NP_001138399.1 | |
NM_001144928.1 | 179 | Silent Mutation | ACA,ACG | T,T 38 | NP_001138400.1 | |
NM_001144929.1 | 179 | Silent Mutation | ACA,ACG | T,T 38 | NP_001138401.1 | |
NM_017595.5 | 179 | Silent Mutation | ACA,ACG | T,T 38 | NP_060065.2 | |
XM_005257251.1 | 179 | Silent Mutation | ACA,ACG | T,T 38 | XP_005257308.1 |
ZNF385C - zinc finger protein 385C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |