Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCACAGTGACCCTGGAGAATCTCA[C/T]AGAGGAGGATGCAGGCACCTACTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606790 | ||||||||||||||||||||
Literature Links: |
CD300A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD300A - CD300a molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256841.1 | 625 | Intron | NP_001243770.1 | |||
NM_007261.3 | 625 | Missense Mutation | ACA,ATA | T,I 94 | NP_009192.2 | |
XM_005256990.2 | 625 | Missense Mutation | ACA,ATA | T,I 94 | XP_005257047.1 | |
XM_005256991.4 | 625 | Missense Mutation | ACA,ATA | T,I 94 | XP_005257048.1 | |
XM_005256992.4 | 625 | UTR 5 | XP_005257049.1 | |||
XM_006721656.3 | 625 | UTR 5 | XP_006721719.1 | |||
XM_011524254.2 | 625 | Intron | XP_011522556.1 |