Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATCCCAGCATTTATCTCCTAAA[C/T]GACCCAGCTCTGTAGATACATAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603696 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C17orf47 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C17orf47 - chromosome 17 open reading frame 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001038704.2 | 1802 | Silent Mutation | TCA,TCG | S,S 567 | NP_001033793.2 | |
XM_011524637.2 | 1802 | Missense Mutation | CAT,CGT | H,R 539 | XP_011522939.1 | |
XM_017024498.1 | 1802 | Intron | XP_016879987.1 |
SEPT4 - septin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SEPT4-AS1 - SEPT4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |