Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCCGACCTCTCGGCTGGAGCTT[C/T]TGTTTGGCATGACCCCGTGTCTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616159 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHRS11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHRS11 - dehydrogenase/reductase 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRM1 - mitochondrial rRNA methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024864.4 | 384 | Silent Mutation | CTG,TTG | L,L 52 | NP_079140.2 | |
XM_005257694.4 | 384 | Silent Mutation | CTG,TTG | L,L 52 | XP_005257751.1 | |
XM_011525275.2 | 384 | Silent Mutation | CTG,TTG | L,L 52 | XP_011523577.1 | |
XM_011525276.2 | 384 | Silent Mutation | CTG,TTG | L,L 52 | XP_011523578.1 | |
XM_017025112.1 | 384 | Silent Mutation | CTG,TTG | L,L 52 | XP_016880601.1 |