Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACGCTGGTTGATGAACTGGATG[A/G]CCTCCTCCAGGCTGCGCACGCACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 100660 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALDH3A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALDH3A1 - aldehyde dehydrogenase 3 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000691.4 | 996 | Missense Mutation | GCC,GTC | A,V 351 | NP_000682.3 | |
NM_001135167.1 | 996 | Missense Mutation | GCC,GTC | A,V 351 | NP_001128639.1 | |
NM_001135168.1 | 996 | Missense Mutation | GCC,GTC | A,V 351 | NP_001128640.1 | |
XM_005256522.1 | 996 | Missense Mutation | GCC,GTC | A,V 468 | XP_005256579.1 | |
XM_005256523.2 | 996 | Missense Mutation | GCC,GTC | A,V 468 | XP_005256580.1 | |
XM_005256524.3 | 996 | Missense Mutation | GCC,GTC | A,V 468 | XP_005256581.1 | |
XM_011523730.1 | 996 | Missense Mutation | GCC,GTC | A,V 278 | XP_011522032.1 | |
XM_011523731.1 | 996 | Missense Mutation | GCC,GTC | A,V 278 | XP_011522033.1 |