Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGGCAGCGCGGGCCCAGCCCCAGA[C/T]GGCAGGGATCCGGGTGCAGCGCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 100725 MIM: 609426 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C17orf107 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C17orf107 - chromosome 17 open reading frame 107 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145536.1 | 452 | Silent Mutation | GAC,GAT | D,D 114 | NP_001139008.1 |
CHRNE - cholinergic receptor nicotinic epsilon subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000080.3 | 452 | Intron | NP_000071.1 | |||
XM_017024115.1 | 452 | Intron | XP_016879604.1 |
MINK1 - misshapen like kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |