Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTACGGATTCCAAACCAAAAGAG[G/T]CAAGCAAGTCTGCGCTGACCCCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610757 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCL4L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCL4L2 - C-C motif chemokine ligand 4 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291468.1 | 879 | Missense Mutation | GGC,GTC | G,V 65 | NP_001278397.1 | |
NM_001291469.1 | 879 | UTR 3 | NP_001278398.1 | |||
NM_001291470.1 | 879 | UTR 3 | NP_001278399.1 | |||
NM_001291471.1 | 879 | UTR 3 | NP_001278400.1 | |||
NM_001291472.1 | 879 | UTR 3 | NP_001278401.1 | |||
NM_001291473.1 | 879 | UTR 3 | NP_001278402.1 | |||
NM_001291474.1 | 879 | UTR 3 | NP_001278403.1 | |||
NM_001291475.1 | 879 | UTR 3 | NP_001278404.1 |