Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGCTGAGGATGAAGCCCGAGCGA[C/T]TCTTGGTGAAGTCGGGCTGGGGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603460 MIM: 606539 | ||||||||||||||||||||
Literature Links: |
CDK5R1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDK5R1 - cyclin dependent kinase 5 regulatory subunit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYO1D - myosin ID | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303279.1 | 3285 | Intron | NP_001290208.1 | |||
NM_001303280.1 | 3285 | Intron | NP_001290209.1 | |||
NM_015194.2 | 3285 | Missense Mutation | AAT,AGT | N,S 995 | NP_056009.1 | |
XM_017024685.1 | 3285 | Intron | XP_016880174.1 |