Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTTCTCGAAATGCTTCATCCCC[A/G]ACAAAGCAAATTTCATGTCCGTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLOD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLOD4 - glyoxalase domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016080.3 | 866 | Silent Mutation | GTC,GTT | V,V 256 | NP_057164.3 | |
XM_006721540.2 | 866 | Silent Mutation | GTC,GTT | V,V 247 | XP_006721603.1 | |
XM_017024711.1 | 866 | Silent Mutation | GTC,GTT | V,V 247 | XP_016880200.1 | |
XM_017024712.1 | 866 | Silent Mutation | GTC,GTT | V,V 200 | XP_016880201.1 |