Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCTCCTGGGGCACGAACTCCTC[C/T]GCAGGCGTGAAATCTTCCCCCCGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137250 MIM: 600947 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAST PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GAST - gastrin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HAP1 - huntingtin associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079870.1 | 1452 | Silent Mutation | GCA,GCG | A,A 488 | NP_001073339.1 | |
NM_001079871.1 | 1452 | Silent Mutation | GCA,GCG | A,A 480 | NP_001073340.1 | |
NM_177977.2 | 1452 | Silent Mutation | GCA,GCG | A,A 505 | NP_817084.2 |