Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATTCTCATTTTACAGAGGAGGAA[A/T]CTGTGCCAGAAGTGGGATCCAGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MILR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MILR1 - mast cell immunoglobulin like receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085423.1 | 916 | Missense Mutation | ACT,TCT | T,S 288 | NP_001078892.1 | |
NM_001291316.1 | 916 | Missense Mutation | ACT,TCT | T,S 193 | NP_001278245.1 | |
NM_001291317.1 | 916 | Missense Mutation | ACT,TCT | T,S 198 | NP_001278246.1 | |
XM_017024486.1 | 916 | Missense Mutation | ACT,TCT | T,S 330 | XP_016879975.1 | |
XM_017024487.1 | 916 | Missense Mutation | ACT,TCT | T,S 230 | XP_016879976.1 |