Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATAATATGATAAATGCCTTAGTA[G/T]AGCTATGCATAAGAGACTATGGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616317 | ||||||||||||||||||||
Literature Links: |
GDPD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GDPD1 - glycerophosphodiester phosphodiesterase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165993.1 | Intron | NP_001159465.1 | ||||
NM_001165994.1 | Intron | NP_001159466.1 | ||||
NM_182569.3 | Intron | NP_872375.2 | ||||
XM_017024521.1 | Intron | XP_016880010.1 |