Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACGGAACCCAGCCGTCTGGTTAG[A/G]GAGAGCTCCGGGGGAGAGGTCCGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 155120 MIM: 611661 | ||||||||||||||||||||
Literature Links: |
ADAM11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAM11 - ADAM metallopeptidase domain 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318933.1 | 104 | UTR 5 | NP_001305862.1 | |||
NM_002390.5 | 104 | Silent Mutation | AGA,AGG | R,R 55 | NP_002381.2 | |
XM_005257373.3 | 104 | Silent Mutation | AGA,AGG | R,R 55 | XP_005257430.1 | |
XM_017024647.1 | 104 | UTR 5 | XP_016880136.1 |
DBF4B - DBF4 zinc finger B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |