Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTAAAAGCCCTGCCTCTCAGCCC[C/T]AGGAAACGTCTGGGTAAACCATCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602627 MIM: 609692 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDC6 - cell division cycle 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001254.3 | 580 | Silent Mutation | CCC,CCT | P,P 55 | NP_001245.1 | |
XM_011525541.1 | 580 | Silent Mutation | CCC,CCT | P,P 55 | XP_011523843.1 | |
XM_011525542.1 | 580 | Silent Mutation | CCC,CCT | P,P 55 | XP_011523844.1 |
WIPF2 - WAS/WASL interacting protein family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |