Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCGGAAGAAATCTTCCAGGATGG[A/G]GAAGGGGAAGAAGGGCAGGAGGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601333 | ||||||||||||||||||||
Literature Links: |
SUPT6H PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SUPT6H - SPT6 homolog, histone chaperone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320755.1 | 737 | Silent Mutation | GGA,GGG | G,G 153 | NP_001307684.1 | |
NM_003170.4 | 737 | Silent Mutation | GGA,GGG | G,G 153 | NP_003161.2 | |
XM_017024978.1 | 737 | Silent Mutation | GGA,GGG | G,G 153 | XP_016880467.1 | |
XM_017024979.1 | 737 | Silent Mutation | GGA,GGG | G,G 153 | XP_016880468.1 | |
XM_017024980.1 | 737 | Silent Mutation | GGA,GGG | G,G 30 | XP_016880469.1 |