Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCCCATGTTTACAGATCCTGAGT[C/T]TCTACCACCAAATTCTGCAAAACTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607742 | ||||||||||||||||||||
Literature Links: |
KRT24 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRT24 - keratin 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019016.2 | 1126 | Missense Mutation | AAC,GAC | N,D 465 | NP_061889.2 | |
XM_006721739.3 | 1126 | Missense Mutation | AAC,GAC | N,D 367 | XP_006721802.1 | |
XM_011524460.2 | 1126 | Missense Mutation | AAC,GAC | N,D 362 | XP_011522762.1 | |
XM_017024299.1 | 1126 | Intron | XP_016879788.1 |
LOC105371775 - uncharacterized LOC105371775 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |