Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTGAGCTTGCTGTTGTGGAAGCC[A/G]CTCCCTCTAGACACAGGCAAAGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610705 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD300LB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD300LB - CD300 molecule like family member b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174892.3 | 501 | Missense Mutation | GCG,GTG | A,V 126 | NP_777552.3 | |
XM_005257027.3 | 501 | Missense Mutation | GCG,GTG | A,V 163 | XP_005257084.1 |