Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCTGTTCGTCTTACAGGCCCCGG[C/G]TCTTCGTTAAGGAAGGAGACGTGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607066 | ||||||||||||||||||||
Literature Links: |
SPATA22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPATA22 - spermatogenesis associated 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170695.1 | 2578 | Intron | NP_001164166.1 | |||
NM_001170696.1 | 2578 | Intron | NP_001164167.1 | |||
NM_001170697.1 | 2578 | Intron | NP_001164168.1 | |||
NM_001170698.1 | 2578 | Intron | NP_001164169.1 | |||
NM_001170699.1 | 2578 | Intron | NP_001164170.1 | |||
NM_001321336.1 | 2578 | Intron | NP_001308265.1 | |||
NM_001321337.1 | 2578 | Intron | NP_001308266.1 | |||
NM_032598.4 | 2578 | Intron | NP_115987.2 |
TRPV3 - transient receptor potential cation channel subfamily V member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258205.1 | 2578 | Missense Mutation | GAC,GAG | D,E 752 | NP_001245134.1 | |
NM_145068.3 | 2578 | Missense Mutation | GAC,GAG | D,E 752 | NP_659505.1 |