Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGGGACCTTCTCCACATACTCCT[C/T]CACCAGGCCCCGCTCGCCACCCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138033 MIM: 616514 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCGR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GCGR - glucagon receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCRIP1 - MAPK regulated corepressor interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001093767.2 | 431 | Missense Mutation | AAG,GAG | K,E 64 | NP_001087236.1 | |
NM_001288798.1 | 431 | Missense Mutation | AAG,GAG | K,E 85 | NP_001275727.1 | |
NM_001288799.1 | 431 | Intron | NP_001275728.1 | |||
NM_207368.4 | 431 | Missense Mutation | AAG,GAG | K,E 64 | NP_997251.2 | |
XM_011523571.2 | 431 | Missense Mutation | AAG,GAG | K,E 85 | XP_011521873.1 | |
XM_011523572.2 | 431 | Intron | XP_011521874.1 |