Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTCAGCTGAAACCAGAAGAAGA[A/C]TATGAAGAGGCACAAGGAGAGGCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614515 MIM: 611850 | ||||||||||||||||||||
Literature Links: |
GPR179 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR179 - G protein-coupled receptor 179 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL45 - mitochondrial ribosomal protein L45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278279.2 | 1037 | Missense Mutation | GAA,GAC | E,D 242 | NP_001265208.2 | |
NM_032351.5 | 1037 | Missense Mutation | GAA,GAC | E,D 292 | NP_115727.5 |