Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTGTCATCCAGGCAGACAGTCTG[C/T]GTGAGGAAGGCCGCTGTAAGAGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608344 MIM: 602301 MIM: 604037 | ||||||||||||||||||||
Literature Links: |
HSPB9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HSPB9 - heat shock protein family B (small) member 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KAT2A - lysine acetyltransferase 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB5C - RAB5C, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252039.1 | 359 | Silent Mutation | ACA,ACG | T,T 93 | NP_001238968.1 | |
NM_004583.3 | 359 | Silent Mutation | ACA,ACG | T,T 60 | NP_004574.2 | |
NM_201434.2 | 359 | Silent Mutation | ACA,ACG | T,T 60 | NP_958842.1 | |
XM_011525090.1 | 359 | Silent Mutation | ACA,ACG | T,T 60 | XP_011523392.1 | |
XM_011525091.1 | 359 | Silent Mutation | ACA,ACG | T,T 60 | XP_011523393.1 |