Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGTCAAAGTAGTGCCCGTAGCC[C/T]CGCTGGATGCGGCTGCTCTCCTCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600723 | ||||||||||||||||||||
Literature Links: |
MPP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MPP2 - membrane palmitoylated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278370.1 | 1778 | Silent Mutation | CGA,CGG | R,R 555 | NP_001265299.1 | |
NM_001278371.1 | 1778 | Silent Mutation | CGA,CGG | R,R 499 | NP_001265300.1 | |
NM_001278372.1 | 1778 | Silent Mutation | CGA,CGG | R,R 534 | NP_001265301.1 | |
NM_001278373.1 | 1778 | Silent Mutation | CGA,CGG | R,R 499 | NP_001265302.1 | |
NM_001278374.1 | 1778 | Silent Mutation | CGA,CGG | R,R 371 | NP_001265303.1 | |
NM_001278375.1 | 1778 | Silent Mutation | CGA,CGG | R,R 499 | NP_001265304.1 | |
NM_001278376.2 | 1778 | Silent Mutation | CGA,CGG | R,R 527 | NP_001265305.2 | |
NM_001278381.1 | 1778 | Silent Mutation | CGA,CGG | R,R 510 | NP_001265310.1 | |
NM_005374.4 | 1778 | Silent Mutation | CGA,CGG | R,R 510 | NP_005365.4 | |
XM_011524827.2 | 1778 | Silent Mutation | CGA,CGG | R,R 588 | XP_011523129.2 |