Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTCAGACTGAGGTGACCCTGGG[C/T]GGTACCATGGGCCAGGCCTCGGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609131 MIM: 610684 MIM: 615019 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLDN7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLDN7 - claudin 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CTDNEP1 - CTD nuclear envelope phosphatase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ELP5 - elongator acetyltransferase complex subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015362.4 | 787 | Silent Mutation | GGC,GGT | G,G 191 | NP_056177.3 | |
NM_203413.2 | 787 | Silent Mutation | GGC,GGT | G,G 191 | NP_981958.1 | |
NM_203414.2 | 787 | Silent Mutation | GGC,GGT | G,G 191 | NP_981959.1 | |
NM_203415.1 | 787 | Silent Mutation | GGC,GGT | G,G 191 | NP_981960.1 | |
XM_011523779.2 | 787 | Silent Mutation | GGC,GGT | G,G 191 | XP_011522081.1 |