Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTCCCACCTGCTCTGTGGCCTCC[A/G]TCCCCATCTCTGGCTACACAGGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600240 MIM: 602346 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCR10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCR10 - C-C motif chemokine receptor 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016602.2 | 12 | Missense Mutation | ACG,ATG | T,M 3 | NP_057686.2 |
CNTNAP1 - contactin associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003632.2 | 12 | Intron | NP_003623.1 | |||
XM_005257748.4 | 12 | Intron | XP_005257805.1 | |||
XM_017025238.1 | 12 | Intron | XP_016880727.1 |
PLEKHH3 - pleckstrin homology, MyTH4 and FERM domain containing H3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |