Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATACCCTGGGAGACCAGGCTGGC[A/G]AGGTTGCTGGTGAGGTTGGATCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616498 MIM: 602976 MIM: 608665 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM134C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM134C - family with sequence similarity 134 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178126.3 | 1047 | Silent Mutation | CTC,CTT | L,L 397 | NP_835227.1 | |
XM_011524440.2 | 1047 | Silent Mutation | CTC,CTT | L,L 300 | XP_011522742.1 |
MLX - MLX, MAX dimerization protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMC3IP - PSMC3 interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |