Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAGAGCTCCCATGAGACAGAGT[C/T]GAAGGGGGCCTCCTTGGACCCGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612749 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPACA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPACA3 - sperm acrosome associated 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317225.1 | Intron | NP_001304154.1 | ||||
NM_001317226.1 | Intron | NP_001304155.1 | ||||
NM_173847.4 | Intron | NP_776246.1 |