Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCAGTTCAAGCTACCCCCGTTG[A/G]CAGCTGTGGTGGCCCCACTGGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611933 MIM: 600328 MIM: 600346 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CISD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CISD3 - CDGSH iron sulfur domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136498.1 | 2200 | UTR 3 | NP_001129970.1 |
MLLT6 - MLLT6, PHD finger domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCGF2 - polycomb group ring finger 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007144.2 | 2200 | Missense Mutation | GCC,GTC | A,V 313 | NP_009075.1 | |
XM_005257640.2 | 2200 | Missense Mutation | GCC,GTC | A,V 313 | XP_005257697.1 | |
XM_005257641.4 | 2200 | Missense Mutation | GCC,GTC | A,V 313 | XP_005257698.1 | |
XM_005257642.3 | 2200 | Missense Mutation | GCC,GTC | A,V 313 | XP_005257699.1 | |
XM_017025016.1 | 2200 | Missense Mutation | GCC,GTC | A,V 313 | XP_016880505.1 |