Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGATTATTTTAGAAAGACTTCA[C/T]CCACAAATGAGAAGACACAATTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609534 MIM: 614853 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATAD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATAD5 - ATPase family, AAA domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024857.4 | 610 | Missense Mutation | CCC,TCC | P,S 71 | NP_079133.3 | |
XM_011525269.2 | 610 | Missense Mutation | CCC,TCC | P,S 71 | XP_011523571.1 | |
XM_011525270.2 | 610 | Missense Mutation | CCC,TCC | P,S 71 | XP_011523572.1 | |
XM_011525273.2 | 610 | Missense Mutation | CCC,TCC | P,S 71 | XP_011523575.1 | |
XM_011525274.2 | 610 | Intron | XP_011523576.1 | |||
XM_017025111.1 | 610 | Missense Mutation | CCC,TCC | P,S 57 | XP_016880600.1 |
CRLF3 - cytokine receptor like factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |