Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCGCCCAGTCTCCCTCTATTGGT[A/G]CACTATCAATTGTGCAGTGCCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611301 MIM: 606590 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAAP100 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAAP100 - Fanconi anemia core complex associated protein 100 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPLOC4 - NPL4 homolog, ubiquitin recognition factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017921.3 | 2578 | UTR 3 | NP_060391.2 | |||
XM_011524979.1 | 2578 | Intron | XP_011523281.1 | |||
XM_011524980.1 | 2578 | UTR 3 | XP_011523282.1 | |||
XM_011524981.1 | 2578 | UTR 3 | XP_011523283.1 | |||
XM_011524982.2 | 2578 | Intron | XP_011523284.1 | |||
XM_017024830.1 | 2578 | UTR 3 | XP_016880319.1 |