Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGAGTGACAGGTGCGAAGTAGGC[A/G]TGGACTTTAATATTGGGTAAATGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609086 | ||||||||||||||||||||
Literature Links: |
FBXL20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXL20 - F-box and leucine rich repeat protein 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184906.1 | 1484 | Silent Mutation | CAC,CAT | H,H 378 | NP_001171835.1 | |
NM_032875.2 | 1484 | Silent Mutation | CAC,CAT | H,H 410 | NP_116264.2 | |
XM_005257746.3 | 1484 | Silent Mutation | CAC,CAT | H,H 447 | XP_005257803.2 | |
XM_005257747.3 | 1484 | Silent Mutation | CAC,CAT | H,H 415 | XP_005257804.2 |